Hafnium dichloride oxide octahydrate, 98+% (metals basis excluding Zr), Zr <1.5%, Thermo Scientific Chemicals
Hafnium dichloride oxide octahydrate, 98+% (metals basis excluding Zr), Zr &lt;1.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium dichloride oxide octahydrate, 98+% (metals basis excluding Zr), Zr <1.5%, Thermo Scientific Chemicals

For Synthesis of Hafnium (IV) Oxide
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011833.22
also known as 011833-22
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS14456-34-9
IUPAC Namedichloro(oxo)hafnium octahydrate
Molecular FormulaCl2H16HfO9
InChI KeyFXQGUVLIMYOGDZ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.Cl[Hf](Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Zr1.9% max.
Impurity contentZr <1.5%
Hafnium dichloride oxide octahydrate is used in the synthesis of hafnium(IV) oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only