Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr &lt;2.7%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011834.22
also known as 011834-22
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
233.00 
Save 34.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
233.00 
Save 34.35 (15%)
Add to cart
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals
Catalog number011834.22
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
233.00 
Save 34.35 (15%)
-
Add to cart
Chemical Identifiers
CAS13499-05-3
IUPAC Namehafnium(4+) tetrachloride
Molecular FormulaCl4Hf
InChI KeyPDPJQWYGJJBYLF-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Hf+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentZr <2.7%
Total Metal Impurities excluding Zr1.9% max.
Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only