Thermo Scientific Chemicals

Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals

Catalog number: 011834.88
2.5 g, Each
Thermo Scientific Chemicals

Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals

Catalog number: 011834.88
2.5 g, Each
Quantity
Catalog number: 011834.88
also known as 011834-88
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13499-05-3
IUPAC Name
hafnium(4+) tetrachloride
Molecular Formula
Cl4Hf
InChI Key
PDPJQWYGJJBYLF-UHFFFAOYSA-J
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Hf+4]
Impurity content
Zr <2.7%
Total Metal Impurities excluding Zr
1.9% max.

Description

Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text