Hafnium(IV) oxide, 99% (metals basis excluding Zr), Zr <1.5%, Thermo Scientific Chemicals
Hafnium(IV) oxide, 99% (metals basis excluding Zr), Zr &lt;1.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium(IV) oxide, 99% (metals basis excluding Zr), Zr <1.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011835.14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS12055-23-1
IUPAC Namehafnium(4+) dioxidandiide
Molecular FormulaHfO2
InChI KeyWIHZLLGSGQNAGK-UHFFFAOYSA-N
SMILES[O--].[O--].[Hf+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Impurity contentZr<1.5%
Total Metal Impurities excluding Zr≤1%
Particle Size-325 mesh
FormPowder
Hafnium(IV) oxide is used in optical coatings and in advanced metal-oxide-semiconductor devices. It is used as a gate insulator in field-effect transistors due to its high dielectric constant. It plays an important role as a possible candidate for resistive-switching memories, as a refractory material in the insulation of devices such as thermocouples and as a high-κ dielectric in DRAM capacitors. Also, used in the preparation of hafnium tetrachloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only