Thermo Scientific Chemicals

Hafnium(IV) oxide, Spectrographic Grade, 99.9% (metals basis excluding Zr), Zr typically <80ppm, Thermo Scientific Chemicals

Catalog number: 011836.04
2 g, Each
Thermo Scientific Chemicals

Hafnium(IV) oxide, Spectrographic Grade, 99.9% (metals basis excluding Zr), Zr typically <80ppm, Thermo Scientific Chemicals

Catalog number: 011836.04
2 g, Each
Quantity
Catalog number: 011836.04
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12055-23-1
IUPAC Name
hafnium(4+) dioxidandiide
Molecular Formula
HfO2
InChI Key
WIHZLLGSGQNAGK-UHFFFAOYSA-N
SMILES
[O--].[O--].[Hf+4]
Total Metal Impurities excluding Zr
0.1% max.

Description

Hafnium(IV) oxide is used in optical coatings and in advanced metal-oxide-semiconductor devices. It is used as a gate insulator in field-effect transistors due to its high dielectric constant. It plays an important role as a possible candidate for resistive-switching memories, as a refractory material in the insulation of devices such as thermocouples and as a high-κ dielectric in DRAM capacitors. Also, used in the preparation of hafnium tetrachloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text