Thallium(I) chloride, 99.999% (metals basis), Thermo Scientific Chemicals
Thallium(I) chloride, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thallium(I) chloride, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 011839.22
also known as 011839-22
Price (USD)/ Each
1,055.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
1,055.00
Add to cart
Thallium(I) chloride, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number011839.22
Price (USD)/ Each
1,055.00
-
Add to cart
Chemical Identifiers
CAS7791-12-0
IUPAC Nameλ¹-thallanylium chloride
Molecular FormulaClTl
InChI KeyGBECUEIQVRDUKB-UHFFFAOYSA-M
SMILES[Cl-].[Tl+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only