Thermo Scientific Chemicals

Phosphorus(V) chloride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011849.22
100 g, Each
Thermo Scientific Chemicals

Phosphorus(V) chloride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011849.22
100 g, Each
Quantity
Catalog number: 011849.22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10026-13-8
IUPAC Name
pentachloro-λ⁵-phosphane
Molecular Formula
Cl5P
InChI Key
UHZYTMXLRWXGPK-UHFFFAOYSA-N
SMILES
ClP(Cl)(Cl)(Cl)Cl
Appearance (Color)
Yellow to yellow-green
Form
Powder
Total Metal Impurities
≤0.01%

Description

Phosphorus(V) chloride is used in organic synthesis, chemical reagents and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text