Thermo Scientific Chemicals

Indium(III) iodide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 011854.04
2 g, Each
Thermo Scientific Chemicals

Indium(III) iodide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 011854.04
2 g, Each
Quantity
Catalog number: 011854.04
also known as 011854-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13510-35-5
IUPAC Name
indium(3+) triiodide
Molecular Formula
I3In
InChI Key
RMUKCGUDVKEQPL-UHFFFAOYSA-K
SMILES
[In+3].[I-].[I-].[I-]
Appearance (Color)
Yellow to red
Total Metal Impurities
≤0.001% (10 ppm)
Form
Crystalline

Description

Indium(III) iodide is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text