Indium(III) iodide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Indium(III) iodide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium(III) iodide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
50 g
Catalog number 011854.09
Price (USD)/ Each
248.65
Online exclusive
276.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
248.65
Online exclusive
276.00 
Save 27.35 (10%)
Add to cart
Indium(III) iodide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number011854.09
Price (USD)/ Each
248.65
Online exclusive
276.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13510-35-5
IUPAC Nameindium(3+) triiodide
Molecular FormulaI3In
InChI KeyRMUKCGUDVKEQPL-UHFFFAOYSA-K
SMILES[In+3].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to red
Total Metal Impurities≤0.001% (10 ppm)
FormCrystalline
Indium(III) iodide is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only