Thermo Scientific Chemicals

Indium(III) hydroxide, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 011855.09
10 g, Each
Thermo Scientific Chemicals

Indium(III) hydroxide, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 011855.09
10 g, Each
Quantity
Catalog number: 011855.09
also known as 011855-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20661-21-6
IUPAC Name
indiganetriol
Molecular Formula
H6InO3
InChI Key
ZMFWDTJZHRDHNW-UHFFFAOYSA-N
SMILES
O.O.O.[In]
Total Metal Impurities
0.002% max.

Description

Indium(III) hydroxide, acts as a precursor to indium(III) oxide. It is used in batteries, glasses, ceramics, chemical reagents, semiconductor materials. It is also used used for manufacturing of panel displays and demist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text