Thermo Scientific Chemicals

Indium(III) chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011859.30
250 g, Each
Thermo Scientific Chemicals

Indium(III) chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011859.30
250 g, Each
Quantity
Catalog number: 011859.30
also known as 011859-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143983-91-9
IUPAC Name
indium(3+) trichloride
Molecular Formula
Cl3In
InChI Key
PSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[In+3]
Total Metal Impurities
0.01% max.

Description

Indium(III) chloride hydrate is used for mild and selective cleavage of tert-butyldimethylsilyl ethers. It is a Lewis acid, and serves as a catalyst for many organic transformations. It also catalyzes the conversion of aldehydes to gem diacetates by chemoselective process. Lithium indium hydride, an in situ reducing agent, can be prepared from indium trichloride. It catalyzes reactions such as Friedel-Crafts acylations and Diels-Alder reactions. It also catalyzes Knoevenagel condensation between the barbituric acid and the aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text