Thermo Scientific Chemicals

Cadmium acetate dihydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 011865.14
25 g, Each
Thermo Scientific Chemicals

Cadmium acetate dihydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 011865.14
25 g, Each
Quantity
Catalog number: 011865.14
also known as 011865-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5743-04-4
IUPAC Name
cadmium(2+) diacetate dihydrate
Molecular Formula
C4H10CdO6
InChI Key
AUIZLSZEDUYGDE-UHFFFAOYSA-L
SMILES
O.O.[Cd++].CC([O-])=O.CC([O-])=O
Total Metal Impurities
0.001% max.

Description

It is used in dyestuff intermediates, ceramics and producing iridescent effects on porcelains and pottery. It acts as a reagent for determination of S, Se, and Te. It is also used in cadmium electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text