Thermo Scientific Chemicals

Manganese(II) chloride, 97%, Thermo Scientific Chemicals

Catalog number: 011868.A3
2 kg, Each
Thermo Scientific Chemicals

Manganese(II) chloride, 97%, Thermo Scientific Chemicals

Catalog number: 011868.A3
2 kg, Each
Quantity
Catalog number: 011868.A3
also known as 011868-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7773-01-5
IUPAC Name
manganese(2+) dichloride
Molecular Formula
Cl2Mn
InChI Key
GLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Mn++]
Assay (unspecified)
0.002% Fe max
Assay (unspecified)
0.009% Ni max
Assay (unspecified)
96.1% min
Residual water
1.0% max

Description

Manganese(II) chloride, plays an important role in the production of dry cell batteries. It is also used as a starting material to the methylcyclopentadienyl manganese tricarbonyl, which has antiknock property. Further, it is used in the formation of an active Manganese(0) species useful for radical cyclization. It is also used in 31P-NMR to determine the size and lamellarity of phospholipidvesicles reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text