Thermo Scientific Chemicals

Ammonium hexachlororuthenate(IV), Ru 28.4% min, Thermo Scientific Chemicals

Catalog number: 011874.03
1 g, Each
Thermo Scientific Chemicals

Ammonium hexachlororuthenate(IV), Ru 28.4% min, Thermo Scientific Chemicals

Catalog number: 011874.03
1 g, Each
Quantity
Catalog number: 011874.03
also known as 011874-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18746-63-9
IUPAC Name
λ⁴-ruthenium(4+) diammonium hexachloride
Molecular Formula
Cl6H8N2Ru
InChI Key
DHTSBQSPUBOKKI-UHFFFAOYSA-J
SMILES
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+4]
Assay (unspecified)
28.4% Ru min

Description

Ammonium hexachlororuthenate(IV) is used in laboratory chemicals and manufacture of other substances. Ammonium hexachlororuthenate (ACR) is a salt precipitated during a substitution reaction between ammonium and ruthenate dissolved in a strong hydrochloric acid medium. This precipitation reaction is used within the platinum industry as a means of recovering and purifying ruthenium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text