Thermo Scientific Chemicals

Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min, Thermo Scientific Chemicals

Catalog number: 011876.09
10 g, Each
Thermo Scientific Chemicals

Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min, Thermo Scientific Chemicals

Catalog number: 011876.09
10 g, Each
Quantity
Catalog number: 011876.09
also known as 011876-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
339268-21-2
IUPAC Name
tetrapotassium hexacyanorutheniumbis(ylium)
Molecular Formula
C6K4N6Ru
InChI Key
FULSJSKUKUZHLZ-UHFFFAOYSA-N
SMILES
[K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N
Assay (unspecified)
Ru 23.0% min

Description

Potassium hexacyanoruthenate(II) hydrate used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text