Thermo Scientific Chemicals

Potassium perruthenate(VII), 97%, Thermo Scientific Chemicals

Catalog number: 011877.03
1 g, Each
Thermo Scientific Chemicals

Potassium perruthenate(VII), 97%, Thermo Scientific Chemicals

Catalog number: 011877.03
1 g, Each
Quantity
Catalog number: 011877.03
also known as 011877-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10378-50-4
IUPAC Name
4-[4,7-bis(benzyloxy)-1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl]-N,N-bis(propan-2-yl)butanamide
Molecular Formula
C32H36N2O5
InChI Key
PXMOBROHNZPLTM-UHFFFAOYSA-N
SMILES
CC(C)N(C(C)C)C(=O)CCCN1C(=O)C2=C(OCC3=CC=CC=C3)C=CC(OCC3=CC=CC=C3)=C2C1=O
Assay (unspecified)
96.1% min

Description

Potassium perruthenate(VII) is used in the production of carbon nanotubes for flexible solid-state supercapacitors. It is also used as an oxidizing agent in the synthesis of 5-formylcytosine from 5-hydroxymethylcytosine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text