Thermo Scientific Chemicals

Potassium tetrabromopalladate(II), Pd 20.0%, Thermo Scientific Chemicals

Catalog number: 011883.03
1 g, Each
Thermo Scientific Chemicals

Potassium tetrabromopalladate(II), Pd 20.0%, Thermo Scientific Chemicals

Catalog number: 011883.03
1 g, Each
Quantity
Catalog number: 011883.03
also known as 011883-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13826-93-2
Assay (unspecified)
Pd 20.0% min

Description

Potassium tetrabromopalladate(II) Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text