Silver benzoate hydrate, Thermo Scientific Chemicals
Silver benzoate hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver benzoate hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 011896.06
also known as 011896-06
Price (USD)/ Each
33.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
33.30
Add to cart
Silver benzoate hydrate, Thermo Scientific Chemicals
Catalog number011896.06
Price (USD)/ Each
33.30
-
Add to cart
Chemical Identifiers
CAS532-31-0
IUPAC Namesilver(1+) benzoate
Molecular FormulaC7H5AgO2
InChI KeyCLDWGXZGFUNWKB-UHFFFAOYSA-M
SMILES[Ag+].[O-]C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water ContentReport Value (Waters of Hydration)
Assay (unspecified)Silver (Ag): ≥42.0%
FormPowder
Silver benzoate hydrate along with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene (MTBD) is an efficient catalytic system for the reaction of carbon dioxide with various ketones. It was used in the synthesis of triphenyltinbenzoate. It is also used as a catalyst in Wolff rearrangement reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only