Thermo Scientific Chemicals

Silver benzoate hydrate, Thermo Scientific Chemicals

Catalog number: 011896.14
25 g, Each
Thermo Scientific Chemicals

Silver benzoate hydrate, Thermo Scientific Chemicals

Catalog number: 011896.14
25 g, Each
Quantity
Catalog number: 011896.14
also known as 011896-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
532-31-0
IUPAC Name
silver(1+) benzoate
Molecular Formula
C7H5AgO2
InChI Key
CLDWGXZGFUNWKB-UHFFFAOYSA-M
SMILES
[Ag+].[O-]C(=O)C1=CC=CC=C1
Water Content
Report Value (Waters of Hydration)
Assay (unspecified)
Silver (Ag): ≥42.0%
Form
Powder

Description

Silver benzoate hydrate along with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene (MTBD) is an efficient catalytic system for the reaction of carbon dioxide with various ketones. It was used in the synthesis of triphenyltinbenzoate. It is also used as a catalyst in Wolff rearrangement reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text