Thermo Scientific Chemicals

Silver diethyldithiocarbamate, ACS, Thermo Scientific Chemicals

Catalog number: 011898.09
10 g, Each
Thermo Scientific Chemicals

Silver diethyldithiocarbamate, ACS, Thermo Scientific Chemicals

Catalog number: 011898.09
10 g, Each
Quantity
Catalog number: 011898.09
also known as 011898-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1470-61-7
IUPAC Name
silver(1+) (diethylcarbamothioyl)sulfanide
Molecular Formula
C5H10AgNS2
InChI Key
NSVHDIYWJVLAGH-UHFFFAOYSA-M
SMILES
[Ag+].CCN(CC)C([S-])=S
Assay (unspecified)
Suitability for determination of arsenic: Passes Test
Solution Test
Solubility in pyridine: Passes Test

Description

Silver diethyldithiocarbamate cluster plays a useful role for the deposition of acanthite (Ag2S) thin films which is employed in photoelectrochemical applications. Silver diethyldithiocarbamate membrane is used to develop ion selective electrodes for nitrate ion. It is useful in the colorimetric determination of arsenic. It acts as a copper chelator, and is utilized to quantitate copper in biological materials by ESR. It also works as an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text