Silver diethyldithiocarbamate, ACS, Thermo Scientific Chemicals
Silver diethyldithiocarbamate, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver diethyldithiocarbamate, ACS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 011898.18
also known as 011898-18
Price (USD)/ Each
865.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
865.00
Add to cart
Silver diethyldithiocarbamate, ACS, Thermo Scientific Chemicals
Catalog number011898.18
Price (USD)/ Each
865.00
-
Add to cart
Chemical Identifiers
CAS1470-61-7
IUPAC Namesilver(1+) (diethylcarbamothioyl)sulfanide
Molecular FormulaC5H10AgNS2
InChI KeyNSVHDIYWJVLAGH-UHFFFAOYSA-M
SMILES[Ag+].CCN(CC)C([S-])=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)greenish-yellow
Assay (unspecified)Suitability for determination of arsenic: Passes Test
Solution TestSolubility in pyridine: Passes Test
Silver diethyldithiocarbamate cluster plays a useful role for the deposition of acanthite (Ag2S) thin films which is employed in photoelectrochemical applications. Silver diethyldithiocarbamate membrane is used to develop ion selective electrodes for nitrate ion. It is useful in the colorimetric determination of arsenic. It acts as a copper chelator, and is utilized to quantitate copper in biological materials by ESR. It also works as an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only