Lead wire, 0.5mm (0.02in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Lead wire, 0.5mm (0.02in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead wire, 0.5mm (0.02in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Form: approx. 2.23g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
Catalog number 011902.G5
also known as 011902-G5
Price (USD)/ Each
177.65
Online exclusive
197.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (USD)/ Each
177.65
Online exclusive
197.00 
Save 19.35 (10%)
Add to cart
Lead wire, 0.5mm (0.02in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number011902.G5
Price (USD)/ Each
177.65
Online exclusive
197.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.5mm (0.02in) diameter.
Appearance (Color)Gray
Assay from Supplier's CofA≥99.998% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only