Tin wire, 0.25mm (0.01in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Tin wire, 0.25mm (0.01in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin wire, 0.25mm (0.01in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Form: Approx. 0.36g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 m
1 m
5 m
Catalog number 011907.G9
also known as 011907-G9
Price (USD)/ Each
396.00
-
Add to cart
Quantity:
25 m
Request bulk or custom format
Price (USD)/ Each
396.00
Add to cart
Tin wire, 0.25mm (0.01in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number011907.G9
Price (USD)/ Each
396.00
-
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormWire, 0.25mm (0.01in) diameter.
Assay from Supplier's CofA≥99.998% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only