Cadmium wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Cadmium wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011909.G5
also known as 011909-G5
Price (USD)/ Each
394.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
464.00 
Save 69.35 (15%)
-
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (USD)/ Each
394.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
464.00 
Save 69.35 (15%)
Add to cart
Cadmium wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number011909.G5
Price (USD)/ Each
394.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
464.00 
Save 69.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-43-9
IUPAC Namecadmium
Molecular FormulaCd
InChI KeyBDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES[Cd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only