Iridium sponge, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Iridium sponge, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iridium sponge, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 012069.04
also known as 012069-04
Price (USD)/ Each
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS7439-88-5
IUPAC Nameiridium
Molecular FormulaIr
InChI KeyGKOZUEZYRPOHIO-UHFFFAOYSA-N
SMILES[Ir]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.05%
FormSolid, -100 mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only