Platinum black, Thermo Scientific Chemicals
Platinum black, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum black, Thermo Scientific Chemicals

Form: S.A. 10m2/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
0.1 g
1 g
Catalog number 012076.06
also known as 012076-06
Price (USD)/ Each
833.65
Online exclusive
926.00 
Save 92.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
833.65
Online exclusive
926.00 
Save 92.35 (10%)
Add to cart
Platinum black, Thermo Scientific Chemicals
Catalog number012076.06
Price (USD)/ Each
833.65
Online exclusive
926.00 
Save 92.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Surface Area10m²/g
Platinum black is used in polymer electrolyte fuel cells, microbial fuel cells, and phosphoric acid fuel cell applications. It is also used as a recombination catalyst for hydrogen elimination. It is also used as a building block for sensor elements. It is used as a catalyst and as a gas absorbent. It is also useful for the electrochemical detection of hydrogen peroxide and nitrite in microchannel. It is useful for the anodic oxidation of ammonia in aqueous potassium hydroxide electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only