Spectroflux 110, Lithium tetraborate & Lithium metaborate, 66.5:33.5 w/w%, Thermo Scientific Chemicals
Spectroflux 110, Lithium tetraborate & Lithium metaborate, 66.5:33.5 w/w%, Thermo Scientific Chemicals
Actual product may vary
Thermo Scientific Chemicals
Spectroflux 110, Lithium tetraborate & Lithium metaborate, 66.5:33.5 w/w%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
012086.A1
also known as 012086-A1
1 kg
012086.D9
also known as 012086-D9
5 x 1 kg
2 Options
Catalog number 012086.A1
also known as 012086-A1
Price (USD)/ Each
478.65
Online exclusive
532.00 
Save 53.35 (10%)
-
Add to cart
Quantity:
1 kg
Price (USD)/ Each
478.65
Online exclusive
532.00 
Save 53.35 (10%)
Add to cart
Spectroflux 110, Lithium tetraborate & Lithium metaborate, 66.5:33.5 w/w%, Thermo Scientific Chemicals
Catalog number012086.A1
Price (USD)/ Each
478.65
Online exclusive
532.00 
Save 53.35 (10%)
-
Add to cart
Intermediate activity flux. Suitable for aluminosilicates and calcareous refractories. Often used in testing of cement and steel. Samples must be fully oxidized. Used to dissolve aluminosilicates ranging from 100% silicon dioxide to 100% aluminum(III) oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Intermediate activity flux. Suitable for aluminosilicates and calcareous refractories. Often used in testing of cement and steel. Samples must be fully oxidized
Specifications
Melting Point875°C
Physical FormSolid
Quantity1 kg
Unit SizeEach