Thermo Scientific™

Osmium(VIII) oxide, 99.8% (metals basis), Os 74.4% min, Thermo Scientific Chemicals

Catalog number: 012103.48
10 x 1 g, Each
Thermo Scientific™

Osmium(VIII) oxide, 99.8% (metals basis), Os 74.4% min, Thermo Scientific Chemicals

Catalog number: 012103.48
10 x 1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 012103.48
also known as 012103-48
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20816-12-0
IUPAC Name
tetraoxoosmium
Molecular Formula
O4Os
InChI Key
VUVGYHUDAICLFK-UHFFFAOYSA-N
SMILES
O=[Os](=O)(=O)=O
Appearance (Color)
Pale yellow
Form
Crystalline solid
Assay (unspecified)
≥74.4% (Osmium)
Total Metal Impurities
≤1%

Description

Oxidation, catalyst in Sharpless dihydroxylations Osmium(VIII) oxide is used in the Sharpless oxyamination to give vicinal amino-alcohols. It is used to oxidize alkenes to the vicinal diols. It acts as a staining agent used in transmission electron microscopy to provide contrast to the image and as a stain for lipids in optical microscopy. As a polymer stain, it is also used to stain selectively copolymers especially styrene-butadiene block copolymers. Further, it is involved in the synthesis of Isosteviol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text