Silver sulfate, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Silver sulfate, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver sulfate, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 012112.14
also known as 012112-14
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
Add to cart
Silver sulfate, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number012112.14
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10294-26-5
IUPAC Namedisilver(1+) sulfuric acid
Molecular FormulaAg2H2O4S
InChI KeyYPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES[Ag+].[Ag+].OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Silver(I) sulfate is employed in silver plating, and in electrolysis. It is a useful non-staining substitute to silver nitrate. Silver sulfate/iodine is an excellent iodination reagent for alkyl, alkoxybenzenes, aromatic amines and uridines. It is used in batteries; its pharmaceutical composition is used as an antimicrobial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only