Thermo Scientific Chemicals

Silver sulfate, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012112.22
100 g, Each
Thermo Scientific Chemicals

Silver sulfate, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012112.22
100 g, Each
Quantity
Catalog number: 012112.22
also known as 012112-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-26-5
IUPAC Name
disilver(1+) sulfuric acid
Molecular Formula
Ag2H2O4S
InChI Key
YPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES
[Ag+].[Ag+].OS(O)(=O)=O
Total Metal Impurities
0.001% max.

Description

Silver(I) sulfate is employed in silver plating, and in electrolysis. It is a useful non-staining substitute to silver nitrate. Silver sulfate/iodine is an excellent iodination reagent for alkyl, alkoxybenzenes, aromatic amines and uridines. It is used in batteries; its pharmaceutical composition is used as an antimicrobial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text