Thermo Scientific Chemicals

Cesium carbonate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals

Catalog number: 012117.06
5 g, Each
Thermo Scientific Chemicals

Cesium carbonate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals

Catalog number: 012117.06
5 g, Each
Quantity
Catalog number: 012117.06
also known as 012117-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
534-17-8
IUPAC Name
dicaesium(1+) carbonate
Molecular Formula
CCs2O3
InChI Key
FJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES
[Cs+].[Cs+].[O-]C([O-])=O
Total Metal Impurities
0.006% max.

Description

Stainless Steel gauze is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text