Thermo Scientific Chemicals

Copper(I) cyanide, Thermo Scientific Chemicals

Catalog number: 012135.30
250 g, Each
Thermo Scientific Chemicals

Copper(I) cyanide, Thermo Scientific Chemicals

Catalog number: 012135.30
250 g, Each
Quantity
Catalog number: 012135.30
also known as 012135-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
544-92-3
IUPAC Name
λ¹-copper(1+) iminomethanide
Molecular Formula
CCuN
InChI Key
DOBRDRYODQBAMW-UHFFFAOYSA-N
SMILES
[Cu+].[C-]#N
Assay (unspecified)
90% min

Description

Copper(I) cyanide is commonly used as a polymerization catalyst. The compound is useful in organic synthesis, as a catalyst, reagent in the preparation of nitriles(sandmeyer reaction). It is also used in electroplating copper and iron, silver plating, brass plating, and copper-tin alloy plating. It is used to make phthalocyanine dyes and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text