Thermo Scientific Chemicals

Potassium cyanide, ACS, 96.0% min, Thermo Scientific Chemicals

Catalog number: 012136.A1
1 kg, Each
Thermo Scientific Chemicals

Potassium cyanide, ACS, 96.0% min, Thermo Scientific Chemicals

Catalog number: 012136.A1
1 kg, Each
Quantity
Catalog number: 012136.A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
151-50-8
IUPAC Name
potassium iminomethanide
Molecular Formula
CKN
InChI Key
NNFCIKHAZHQZJG-UHFFFAOYSA-N
SMILES
[K+].[C-]#N
Assay (unspecified)
96.0% KCN min
Impurity content
Cl 0.5%
Impurity content
Iron total (as Fe) 0.03%
Impurity content
Na 0.5%
Impurity content
Pb 2ppm

Description

KCN is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von Richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text