Thermo Scientific Chemicals

Sodium cyanide, ACS, 95% min, Thermo Scientific Chemicals

Catalog number: 012137.36
500 g, Each
Thermo Scientific Chemicals

Sodium cyanide, ACS, 95% min, Thermo Scientific Chemicals

Catalog number: 012137.36
500 g, Each
Quantity
Catalog number: 012137.36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
143-33-9
IUPAC Name
sodium iminomethanide
Molecular Formula
CNNa
InChI Key
MNWBNISUBARLIT-UHFFFAOYSA-N
SMILES
[Na+].[C-]#N
Impurity content
Lead (Pb): ≤5ppm
Impurity content
Sulfate (SO₄): ≤0.05%
Assay from Suppliers CofA
≥95.0% NaCN
Impurity content
Iron total (as Fe): ≤0.005%
Form
Powder or granular powder

Description

Sodium cyanide is used as a starting material for the preparation of Reissert compounds, cyanogen bromide, cyanuric chloride and cyanogen chloride. It acts as a catalyst for the aminolysis of esters to primary amides. Furthermore, it is used for fumigation, in electroplating and for extracting gold and silver in mining industry. In organic synthesis, it is involved in the cyanation reaction of alkyl halides under phase transfer conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text