Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals
Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012140.30
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS12045-64-6
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Hf0.5% max.
Zirconium boride is use to improve resistance in zirconia-based, carbon-bonded refractories in contact with ferrous melts. Used in nuclear applications, aerospace refractory, in cutting tools and to protect thermocouple tubes. It also used in use of its relatively high conductivity, especially for a ceramic.