Thermo Scientific Chemicals

Tantalum carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 012144.09
10 g, Each
Thermo Scientific Chemicals

Tantalum carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 012144.09
10 g, Each
Quantity
Catalog number: 012144.09
also known as 012144-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12070-06-3
IUPAC Name
methanidylidynetantalumylium
Molecular Formula
CTa
InChI Key
DUMHRFXBHXIRTD-UHFFFAOYSA-N
SMILES
[C-]#[Ta+]
Total Metal Impurities
0.5% max.

Description

Tantalum carbides is used in  tungsten carbide. It is also used as a coating for steel moulds in the injection moulding of aluminum alloys. While providing a hard, wear resistant surface, it also provides a low friction mould surface. It is also used in the production of sharp instruments with extreme mechanical resistance and hardness, tool bits for cutting tools.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text