Thermo Scientific Chemicals

Niobium nitride, Thermo Scientific Chemicals

Catalog number: 012146.09
10 g, Each
Thermo Scientific Chemicals

Niobium nitride, Thermo Scientific Chemicals

Catalog number: 012146.09
10 g, Each
Quantity
Catalog number: 012146.09
also known as 012146-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24621-21-4
IUPAC Name
niobioamine
Molecular Formula
NNb
InChI Key
CFJRGWXELQQLSA-UHFFFAOYSA-N
SMILES
N#[Nb]
Assay (unspecified)
99% min

Description

Niobium nitride is used as superconductor. Used in absorbing anti-reflective coatings. It used to produce arc-welding rods and in steel. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text