Thermo Scientific Chemicals

Niobium carbide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 012147.22
100 g, Each
Thermo Scientific Chemicals

Niobium carbide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 012147.22
100 g, Each
Quantity
Catalog number: 012147.22
also known as 012147-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12069-94-2
IUPAC Name
methanidylidyneniobiumylium
Molecular Formula
CNb
InChI Key
AADPBXLVMHIYCT-UHFFFAOYSA-N
SMILES
[C-]#[Nb+]
Total Metal Impurities
0.9% max.

Description

Niobium carbide is used in tool bits for cutting tools.  Zirconium carbide and niobium carbide can be used as refractory coatings in nuclear reactors. It is used to produce arc-welding rods and in corrosion-resistant steel. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text