Sodium tetrachloroaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 49-50%, Thermo Scientific Chemicals
Sodium tetrachloroaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 49-50%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium tetrachloroaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 49-50%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 012148.03
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
Add to cart
Sodium tetrachloroaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 49-50%, Thermo Scientific Chemicals
Catalog number012148.03
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13874-02-7
IUPAC Namesodium tetrachlorogolduide dihydrate
Molecular FormulaAuCl4H4NaO2
InChI KeyOEERDTKOVZFOHF-UHFFFAOYSA-J
SMILESO.O.[Na+].Cl[Au-](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow/orange
FormCrystalline
Assay (special)Au 49-50%
Total Metal Impurities≤0.01%
It is used in chemo-therapeutics. It is employed as a catalyst in the removal of tert-butyl(dimethyl)silyl (TBS) protecting groups. It is selective in the presence of aromatic TBS ethers, aliphatic triisopropylsilyl ethers, aliphatic tert-butyl(diphenyl)silyl ethers, or sterically hindered aliphatic TBS ethers. With excess catalyst, TBS ethers can also be transformed into 4-methoxybenzyl ethers or methyl ethers in one pot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only