Thermo Scientific Chemicals

Potassium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 49% min, Thermo Scientific Chemicals

Catalog number: 012150.03
1 g, Each
Thermo Scientific Chemicals

Potassium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 49% min, Thermo Scientific Chemicals

Catalog number: 012150.03
1 g, Each
Quantity
Catalog number: 012150.03
also known as 012150-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27988-75-6
IUPAC Name
potassium tetrachlorogoldtris(ylium)
Molecular Formula
AuCl4K
InChI Key
IKHCGJWURGSQRT-UHFFFAOYSA-J
SMILES
[K+].Cl[Au+3](Cl)(Cl)Cl
Assay (unspecified)
Au 49% min
Total Metal Impurities
0.01% max.

Description

Potassium tetrachloroaurate(III) hydrate is used as a raw material for the preparation of gold(III) dithiolate complexes, which finds application in luminescence studies, photosensitizers and photocatalysts. Further, it is used for gold electroplating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text