Potassium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 49% min, Thermo Scientific Chemicals
Potassium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 49% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 49% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 012150.06
also known as 012150-06
Price (USD)/ Each
645.65
Online exclusive
717.00 
Save 71.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
645.65
Online exclusive
717.00 
Save 71.35 (10%)
Add to cart
Potassium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 49% min, Thermo Scientific Chemicals
Catalog number012150.06
Price (USD)/ Each
645.65
Online exclusive
717.00 
Save 71.35 (10%)
-
Add to cart
Chemical Identifiers
CAS27988-75-6
IUPAC Namepotassium tetrachlorogoldtris(ylium)
Molecular FormulaAuCl4K
InChI KeyIKHCGJWURGSQRT-UHFFFAOYSA-J
SMILES[K+].Cl[Au+3](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Au 49% min
Total Metal Impurities0.01% max.
Potassium tetrachloroaurate(III) hydrate is used as a raw material for the preparation of gold(III) dithiolate complexes, which finds application in luminescence studies, photosensitizers and photocatalysts. Further, it is used for gold electroplating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only