Chromium silicide, 99+% (metals basis), Thermo Scientific Chemicals
Chromium silicide, 99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium silicide, 99+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 012151.30
also known as 012151-30
Price (USD)/ Each
275.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
275.00
Add to cart
Chromium silicide, 99+% (metals basis), Thermo Scientific Chemicals
Catalog number012151.30
Price (USD)/ Each
275.00
-
Add to cart
Chemical Identifiers
CAS12018-09-6
Molecular FormulaCrSi2
InChI KeyCHXGWONBPAADHP-UHFFFAOYSA-N
SMILES[Si]#[Cr]#[Si]
Molecular Weight (g/mol)108.17
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Chromium silicide is applied in production of ceramic materials. It can also produce an oxidation-resistant surface in a low pressure plasma spraying.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only