Thermo Scientific Chemicals

Gold(III) hydroxide, Au 79% min, Thermo Scientific Chemicals

Catalog number: 012159.03
1 g, Each
Thermo Scientific Chemicals

Gold(III) hydroxide, Au 79% min, Thermo Scientific Chemicals

Catalog number: 012159.03
1 g, Each
Quantity
Catalog number: 012159.03
also known as 012159-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1303-52-2
Assay (unspecified)
Au 79% min

Description

Gold(III) hydroxide is used in medicine, porcelain making, gold plating, and daguerrotypes. Used for preparation of gold catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text