Gold(III) hydroxide, Au 79% min
Gold(III) hydroxide, Au 79% min
Gold(III) hydroxide, Au 79% min
Gold(III) hydroxide, Au 79% min
Thermo Scientific Chemicals

Gold(III) hydroxide, Au 79% min

Gold plating solutions, porcelain decoration | CAS: 1303-52-2 | AuH6O3 | 251.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number 012159.06
also known as 012159-06
Price (USD)
935.65
Special offer
Online exclusive
Ends: 26-Sep-2025
1,156.00
Save 220.35 (19%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
935.65
Special offer
Online exclusive
Ends: 26-Sep-2025
1,156.00
Save 220.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS1303-52-2
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Au 79% min
Gold(III) hydroxide is used in medicine, porcelain making, gold plating, and daguerrotypes. Used for preparation of gold catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Share catalog number, name or link