Thermo Scientific Chemicals

Gold(III) oxide, Premion™, 99.99% (metals basis), Au 88.6% min, Thermo Scientific Chemicals

Catalog number: 012161.03
1 g, Each
Thermo Scientific Chemicals

Gold(III) oxide, Premion™, 99.99% (metals basis), Au 88.6% min, Thermo Scientific Chemicals

Catalog number: 012161.03
1 g, Each
Quantity
Catalog number: 012161.03
also known as 012161-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1303-58-8
IUPAC Name
digold(3+) trioxidandiide
Molecular Formula
Au2O3
InChI Key
DDYSHSNGZNCTKB-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Au+3].[Au+3]
Assay (unspecified)
Au 88.6% min
Total Metal Impurities
0.01% max.

Description

It is used in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity. It acts as strong reducing agent in redox reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text