Thermo Scientific Chemicals

Gold(III) bromide, Premion™, 99.99% (metals basis), Au 44.6% min, Thermo Scientific Chemicals

Catalog number: 012162.03
1 g, Each
Thermo Scientific Chemicals

Gold(III) bromide, Premion™, 99.99% (metals basis), Au 44.6% min, Thermo Scientific Chemicals

Catalog number: 012162.03
1 g, Each
Quantity
Catalog number: 012162.03
also known as 012162-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-28-7
Assay (unspecified)
Au 44.6% min
Total Metal Impurities
0.01% max.

Description

Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text