Thermo Scientific Chemicals

Gold(III) chloride, Au 64.4% min, Thermo Scientific Chemicals

Catalog number: 012163.06
5 g, Each
Thermo Scientific Chemicals

Gold(III) chloride, Au 64.4% min, Thermo Scientific Chemicals

Catalog number: 012163.06
5 g, Each
Quantity
Catalog number: 012163.06
also known as 012163-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13453-07-1
IUPAC Name
gold hydrochloridyl
Molecular Formula
AuCl
InChI Key
BRSVJNYNWNMJKC-UHFFFAOYSA-N
SMILES
[Cl].[Au]
Assay (unspecified)
64.4-65.5% Au

Description

It is used in colloidal gold solutions, in photography and as a print toning agent(gold toning), starting solution to form other gold compounds and a precursor for preparation of ultra pure gold metal. It is used in electroplating and electroless plating as an anode in an electric cell. Gold(III) chloride acts as a gold catalyst and cell body stains for bright field and dark field microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text