Potassium hexachloroplatinate(IV), Pt 39.6%, Thermo Scientific Chemicals
Potassium hexachloroplatinate(IV), Pt 39.6%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hexachloroplatinate(IV), Pt 39.6%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number 012169.01
also known as 012169-01
Price (USD)/ Each
45.50
-
Add to cart
Quantity:
0.25 g
Request bulk or custom format
Price (USD)/ Each
45.50
Add to cart
Potassium hexachloroplatinate(IV), Pt 39.6%, Thermo Scientific Chemicals
Catalog number012169.01
Price (USD)/ Each
45.50
-
Add to cart
Chemical Identifiers
CAS16921-30-5
IUPAC Namedipotassium hexachloroplatinumdiuide
Molecular FormulaCl6K2Pt
InChI KeyDPAIVKJGTXERIM-UHFFFAOYSA-H
SMILES[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)39.6% Pt min
Potassium hexachloroplatinate(IV) is used for the determination of potassium by gravimetric analysis. It is also used to prepare potassium tetrchloroplatinate by reduction using hydrazine dihydrochloride. Further, it is used in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only