Thermo Scientific Chemicals

Potassium hexachloroplatinate(IV), Pt 39.6%, Thermo Scientific Chemicals

Catalog number: 012169.06
5 g, Each
Thermo Scientific Chemicals

Potassium hexachloroplatinate(IV), Pt 39.6%, Thermo Scientific Chemicals

Catalog number: 012169.06
5 g, Each
Quantity
Catalog number: 012169.06
also known as 012169-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16921-30-5
IUPAC Name
dipotassium hexachloroplatinumdiuide
Molecular Formula
Cl6K2Pt
InChI Key
DPAIVKJGTXERIM-UHFFFAOYSA-H
SMILES
[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl
Assay (unspecified)
39.6% Pt min

Description

Potassium hexachloroplatinate(IV) is used for the determination of potassium by gravimetric analysis. It is also used to prepare potassium tetrchloroplatinate by reduction using hydrazine dihydrochloride. Further, it is used in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text