Thermo Scientific Chemicals

Platinum(II) iodide, Premion™, 99.99% (metals basis), Pt 43.0% min, Thermo Scientific Chemicals

Catalog number: 012170.06
5 g, Each
Thermo Scientific Chemicals

Platinum(II) iodide, Premion™, 99.99% (metals basis), Pt 43.0% min, Thermo Scientific Chemicals

Catalog number: 012170.06
5 g, Each
Quantity
Catalog number: 012170.06
also known as 012170-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-39-8
IUPAC Name
platinum(2+) diiodide
Molecular Formula
I2Pt
InChI Key
ZXDJCKVQKCNWEI-UHFFFAOYSA-L
SMILES
[I-].[I-].[Pt++]
Assay (unspecified)
Pt 43.0% min
Total Metal Impurities
0.01% max.

Description

Platinum(II) iodide is used in pharmaceuticals and medical applications. It is also used to form cis and trans complexes with propan-1-amine. It is utilized in the catalytic hydration reaction of alkynes such as 2-hexyne, 4-methyl-2-pentyne, and 4,4dimethyl-2- pentyne . It is also used in the preparation of diiodo(2,9-dimethyl-1,10-phenanthroline)platinum(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text